Assay Details
Target Gene Details
Entrez Gene ID: | 130399 |
Gene Name: | activin A receptor type 1C |
Gene Aliases: |
ACVRLK7, ALK7 |
Location: |
Chr.2:157526767-157628887 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR1C | NM_001111031.1 | 9 | 1550 | NP_001104501.1 |
| NM_001111032.1 | 8 | 1425 | NP_001104502.1 | |
| NM_001111033.1 | 7 | 1194 | NP_001104503.1 | |
| NM_145259.2 | 9 | 1665 | NP_660302.2 | |
| AF525679.1 | 9 | 1553 | AAP21993.1 | |
| AF525680.1 | 8 | 1264 | AAP21994.1 | |
| AF525681.1 | 7 | 1033 | AAP21995.1 | |
| AY127050.1 | 9 | 1504 | AAM93495.1 | |
| BC022530.1 | 9 | 1665 | AAH22530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011505 | Chr.2:157303324 - 157624221 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map