Assay Details
Target Gene Details
Entrez Gene ID: | 55011 |
Gene Name: | PIH1 domain containing 1 |
Gene Aliases: |
NOP17 |
Location: |
Chr.19:49446293-49452332 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIH1D1 | NM_017916.2 | 1 | 75 | NP_060386.1 |
| AK000650.1 | BAA91307.1 | |||
| AK294687.1 | ||||
| AK304476.1 | ||||
| BC001108.2 | AAH01108.1 | |||
| DC385084.1 | 1 | 75 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map