Assay Details
Target Gene Details
Entrez Gene ID: | 11275 |
Gene Name: | kelch like family member 2 |
Gene Aliases: |
ABP-KELCH, MAV, MAYVEN |
Location: |
Chr.4:165207618-165323156 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL2 | NM_001161521.1 | 8 | 995 | NP_001154993.1 |
| NM_001161522.1 | 7 | 888 | NP_001154994.1 | |
| NM_007246.3 | 8 | 1118 | NP_009177.3 | |
| XM_011531572.2 | 9 | 1108 | XP_011529874.1 | |
| XM_011531575.2 | 7 | 940 | XP_011529877.1 | |
| XM_011531576.2 | 6 | 608 | XP_011529878.1 | |
| XM_017007674.1 | 9 | 1068 | XP_016863163.1 | |
| XM_017007675.1 | 7 | 833 | XP_016863164.1 | |
| XM_017007676.1 | 7 | 941 | XP_016863165.1 | |
| XM_017007677.1 | 6 | 815 | XP_016863166.1 | |
| AB209727.1 | 4 | 462 | BAD92964.1 | |
| AF059569.1 | 8 | 967 | AAC67502.1 | |
| AK025289.1 | 5 | 584 | ||
| AK294103.1 | 8 | 989 | ||
| AK294336.1 | 4 | 417 | ||
| AK308940.1 | 4 | 418 | ||
| AK315372.1 | 8 | 963 | ||
| BC022503.1 | 8 | 1100 | AAH22503.1 | |
| BC036468.2 | 8 | 1059 | AAH36468.1 | |
| BX647776.1 | 7 | 872 |
More Information
Set Membership: |
|

Genomic Map