Assay Details
Target Gene Details
Entrez Gene ID: | 1429 |
Gene Name: | crystallin zeta |
Gene Aliases: |
- |
Location: |
Chr.1:74705482-74733408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYZ | NM_001130042.1 | NP_001123514.1 | ||
| NM_001130043.1 | NP_001123515.1 | |||
| NM_001134759.1 | NP_001128231.1 | |||
| NM_001889.3 | NP_001880.2 | |||
| XM_005270491.4 | XP_005270548.1 | |||
| XM_011540747.1 | XP_011539049.1 | |||
| XM_017000367.1 | XP_016855856.1 | |||
| XM_017000368.1 | XP_016855857.1 | |||
| AB209714.1 | BAD92951.1 | |||
| AK128794.1 | ||||
| AK223150.1 | BAD96870.1 | |||
| AK223201.1 | BAD96921.1 | |||
| AK314813.1 | ||||
| BC039578.1 | AAH39578.1 | |||
| BC070058.1 | AAH70058.1 | |||
| BX647883.1 | CAI46072.1 | |||
| CN368109.1 | ||||
| DA256305.1 | ||||
| L13278.1 | AAA36536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586466 | Chr.1:74714333 - 74726895 on Build GRCh38 | Loss |
|
| nsv830259 | Chr.1:74623493 - 74775378 on Build GRCh38 | Gain |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|
| esv2662359 | Chr.1:74714315 - 74726928 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map