Assay Details
Target Gene Details
Entrez Gene ID: | 219790 |
Gene Name: | rhotekin 2 |
Gene Aliases: |
PLEKHK1, bA531F24.1 |
Location: |
Chr.10:62159131-62268879 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTKN2 | NM_145307.3 | 12 | 5447 | NP_660350.2 |
| XM_011539456.2 | 14 | 5568 | XP_011537758.1 | |
| XM_011539457.2 | XP_011537759.1 | |||
| XM_011539460.2 | XP_011537762.1 | |||
| XM_017015842.1 | 13 | 5470 | XP_016871331.1 | |
| XM_017015843.1 | XP_016871332.1 | |||
| XM_017015844.1 | XP_016871333.1 | |||
| AK096671.1 | BAC04837.1 | |||
| AY150309.1 | 12 | 5291 | AAN71738.1 | |
| CR749217.1 | 1 | 3593 | CAH18074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8681 | Chr.10:61896798 - 62671529 on Build GRCh38 | Gain |
|
| nsv831890 | Chr.10:62167783 - 62321747 on Build GRCh38 | Gain |
|
| nsv6831 | Chr.10:62184801 - 62217947 on Build GRCh38 | Insertion |
|
| esv34099 | Chr.10:62021186 - 62427805 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map