Assay Details
Target Gene Details
Entrez Gene ID: | 2882 |
Gene Name: | glutathione peroxidase 7 |
Gene Aliases: |
CL683, GPX6, GPx-7, GSHPx-7, NPGPx |
Location: |
Chr.1:52602371-52609051 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX7 | NM_015696.4 | NP_056511.2 | ||
| AF320068.2 | AAN76501.1 | |||
| AK027683.1 | BAB55294.1 | |||
| AY358402.1 | AAQ88768.1 | |||
| BC032788.1 | AAH32788.1 | |||
| BI550170.1 |
More Information
Set Membership: |
|

Genomic Map