Assay Details
Target Gene Details
Entrez Gene ID: | 285175 |
Gene Name: | unc-80 homolog, NALCN activator |
Gene Aliases: |
C2orf21, UNC-80 |
Location: |
Chr.2:209771925-209999300 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
More Information
Set Membership: |
|

Genomic Map