Assay Details
Target Gene Details
Entrez Gene ID: | 22983 |
Gene Name: | microtubule associated serine/threonine kinase 1 |
Gene Aliases: |
SAST, SAST170 |
Location: |
Chr.19:12833931-12874953 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAST1 | NM_014975.2 | NP_055790.1 | ||
| XM_011527805.2 | XP_011526107.1 | |||
| XM_017026501.1 | XP_016881990.1 | |||
| AB023190.1 | ||||
| AK297553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2420 | Chr.19:12828187 - 12863658 on Build GRCh38 | Insertion |
|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map