Assay Details
Target Gene Details
Entrez Gene ID: | 257202 |
Gene Name: | glutathione peroxidase 6 |
Gene Aliases: |
GPX5p, GPXP3, GPx-6, GSHPx-6, dJ1186N24, dJ1186N24.1 |
Location: |
Chr.6:28503296-28515793 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX6 | NM_182701.1 | 5 | 818 | NP_874360.1 |
| AY324826.1 | 5 | 818 | AAP85543.1 | |
| BC133691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576110 | Chr.6:28449711 - 28681348 on Build GRCh38 | Gain |
|
| nsv601210 | Chr.6:28449445 - 28606638 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map