Assay Details
Target Gene Details
Entrez Gene ID: | 4256 |
Gene Name: | matrix Gla protein |
Gene Aliases: |
GIG36, MGLAP, NTI |
Location: |
Chr.12:14881181-14885919 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGP | NM_000900.3 | NP_000891.2 | ||
| NM_001190839.1 | NP_001177768.1 | |||
| CB108937.1 | ||||
| N85639.1 |
More Information
Set Membership: |
|

Genomic Map