Assay Details
Target Gene Details
Entrez Gene ID: | 84678 |
Gene Name: | lysine demethylase 2B |
Gene Aliases: |
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2 |
Location: |
Chr.12:121402289-121582268 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2B | XM_011538867.2 | XP_011537169.1 |
Target Gene Details
Entrez Gene ID: | 80196 |
Gene Name: | ring finger protein 34 |
Gene Aliases: |
CARP-1, CARP1, RFI, RIF, RIFF, hRFI |
Location: |
Chr.12:121400083-121424353 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF34 | NM_001256858.1 | 5 | 1045 | NP_001243787.1 |
| NM_025126.3 | 6 | 1621 | NP_079402.2 | |
| NM_194271.2 | 7 | 1652 | NP_919247.1 | |
| AF306709.1 | 6 | 1663 | AAK51328.1 | |
| AK024957.1 | 1 | 1574 | ||
| AK025439.1 | 6 | 1586 | BAB15132.1 | |
| AK096994.1 | 3 | 1202 | ||
| AK225285.1 | 6 | 1586 | ||
| BC007826.2 | 6 | 1546 | AAH07826.1 |
More Information
Set Membership: |
|

Genomic Map