Assay Details
Target Gene Details
Entrez Gene ID: | 10648 |
Gene Name: | secretoglobin family 1D member 1 |
Gene Aliases: |
LIPA, LPHA, LPNA |
Location: |
Chr.11:62190238-62193537 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCGB1D1 | NM_006552.1 | NP_006543.1 | ||
| AJ224171.1 | CAA11863.1 | |||
| BC062693.1 | AAH62693.1 | |||
| BC069170.1 | AAH69170.1 | |||
| BC069289.1 | AAH69289.1 |
More Information
Set Membership: |
|

Genomic Map