Assay Details
Target Gene Details
Entrez Gene ID: | 10826 |
Gene Name: | fatty acid hydroxylase domain containing 2 |
Gene Aliases: |
C5orf4 |
Location: |
Chr.5:154817018-154850685 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAXDC2 | XM_017008965.1 | 10 | 4315 | XP_016864454.1 |
Target Gene Details
Entrez Gene ID: | 23367 |
Gene Name: | La ribonucleoprotein domain family member 1 |
Gene Aliases: |
LARP |
Location: |
Chr.5:154682663-154817607 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP1 | NM_015315.4 | NP_056130.2 | ||
| XM_005268404.3 | XP_005268461.1 | |||
| XM_005268406.3 | XP_005268463.1 | |||
| XM_005268407.3 | XP_005268464.1 | |||
| XM_005268408.3 | XP_005268465.1 | |||
| XM_006714775.2 | XP_006714838.1 | |||
| XM_006714776.3 | XP_006714839.1 | |||
| XM_011537614.1 | XP_011535916.1 | |||
| XM_011537615.1 | XP_011535917.1 | |||
| XM_011537616.1 | XP_011535918.1 | |||
| XM_011537617.1 | XP_011535919.1 | |||
| XM_011537618.2 | XP_011535920.1 | |||
| XM_017009283.1 | XP_016864772.1 | |||
| XM_017009284.1 | XP_016864773.1 | |||
| BQ003824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830521 | Chr.5:154768007 - 154864706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map