Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | 42 | 6434 | NP_001252541.1 |
| NM_016284.4 | 42 | 6449 | NP_057368.3 | |
| NR_049763.1 | 42 | 6434 | ||
| AB023224.2 | 29 | 4494 | BAA76851.2 | |
| AF110778.1 | 2 | 226 | AAF14861.1 | |
| AK056927.1 | 19 | 2779 | ||
| AK075125.1 | 12 | 1748 | ||
| AK092885.1 | 2 | 991 | ||
| AK301092.1 | 2 | 213 | ||
| AK302495.1 | 19 | 2819 | ||
| AL117492.1 | 14 | 2288 | CAB55960.1 | |
| AL832769.1 | 42 | 6398 | ||
| AL833549.1 | 42 | 6408 | ||
| BC000779.2 | 4 | 607 | AAH00779.2 | |
| BC024317.1 | 25 | 3848 | AAH24317.1 | |
| BX537840.1 | 42 | 6380 | CAD97851.1 | |
| CR749237.1 | 35 | 5389 | CAH18093.1 | |
| EF553522.1 | 42 | 6383 |
More Information
Set Membership: |
|

Genomic Map