Assay Details
Target Gene Details
Entrez Gene ID: | 100528022 |
Gene Name: | BLOC1S1-RDH5 readthrough |
Gene Aliases: |
- |
Location: |
Chr.12:55716034-55724742 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S1-RDH5 | NR_037658.1 | |||
| AK096656.1 | ||||
| AK307141.1 | ||||
| BG722558.1 |
Target Gene Details
Entrez Gene ID: | 5959 |
Gene Name: | retinol dehydrogenase 5 |
Gene Aliases: |
9cRDH, HSD17B9, RDH1, SDR9C5 |
Location: |
Chr.12:55720367-55724742 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH5 | NM_001199771.1 | NP_001186700.1 | ||
| NM_002905.3 | NP_002896.2 | |||
| AK293313.1 | ||||
| AK307157.1 | ||||
| BC028298.1 | AAH28298.1 | |||
| BI765016.1 | ||||
| BP381700.1 | ||||
| U43559.1 | AAC50725.1 | |||
| U89717.1 | AAB93668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
| nsv559004 | Chr.12:55704254 - 55777109 on Build GRCh38 | Loss |
|
| nsv525231 | Chr.12:55683135 - 55730548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map