Assay Details
Target Gene Details
Entrez Gene ID: | 285195 |
Gene Name: | solute carrier family 9 member A9 |
Gene Aliases: |
AUTS16, NHE9 |
Location: |
Chr.3:143265222-143848531 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A9 | NM_173653.3 | 16 | 2695 | NP_775924.1 |
| XM_011512703.2 | 13 | 2005 | XP_011511005.1 | |
| XM_017006203.1 | 15 | 2453 | XP_016861692.1 | |
| AB075486.1 | 6 | 1196 | BAE45746.1 | |
| AK092932.1 | 10 | 1746 | BAC04005.1 | |
| AK301816.1 | ||||
| AL832304.1 | 16 | 2560 | CAI46158.1 | |
| AY254100.1 | 16 | 2610 | AAP80573.1 | |
| BC035779.1 | 16 | 2611 | AAH35779.1 |
More Information
Set Membership: |
|

Genomic Map