Assay Details
Target Gene Details
Entrez Gene ID: | 285126 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C5 gamma |
Gene Aliases: |
CSP-gamma |
Location: |
Chr.2:27275421-27281430 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC5G | NM_001303127.1 | 7 | 1865 | NP_001290056.1 |
| NM_001303128.1 | 6 | 1665 | NP_001290057.1 | |
| NM_173650.2 | 7 | 1927 | NP_775921.1 | |
| NR_130126.1 | 4 | 1285 | ||
| XM_006711995.3 | 7 | 1839 | XP_006712058.1 | |
| AI183575.1 | 1 | 84 | ||
| AK097736.1 | 7 | 1927 | BAC05155.1 | |
| BC035194.2 | 7 | 1824 | AAH35194.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115076091] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map