Assay Details
Target Gene Details
Entrez Gene ID: | 3087 |
Gene Name: | hematopoietically expressed homeobox |
Gene Aliases: |
HEX, HMPH, HOX11L-PEN, PRH, PRHX |
Location: |
Chr.10:92689924-92695651 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHEX | NM_002729.4 | 2 | 479 | NP_002720.1 |
| AK309463.1 | 2 | 1246 | ||
| AK314891.1 | 2 | 448 | ||
| BC014336.1 | 2 | 452 | AAH14336.1 | |
| BC015110.1 | 2 | 452 | AAH15110.1 | |
| BC050638.1 | 2 | 419 | AAH50638.1 | |
| DB504532.1 | ||||
| L16499.1 | 2 | 451 | AAA02988.1 | |
| X67235.1 | 2 | 423 | CAA47661.1 | |
| Z21533.1 | 1 | 397 | CAA79730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv168n67 | Chr.10:92637392 - 92712752 on Build GRCh38 | Gain |
|
| nsv1046258 | Chr.10:92107212 - 92706349 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map