Assay Details
Target Gene Details
Entrez Gene ID: | 5368 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ |
Location: |
Chr.8:28316979-28343355 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNOC | NM_006228.4 | 2 | 266 | NP_006219.1 |
| XM_005273532.2 | 2 | 353 | XP_005273589.1 | |
| XM_011544559.2 | 2 | 156 | XP_011542861.1 | |
| AK289633.1 | 2 | 129 | ||
| AY335948.1 | 2 | 128 | AAP94601.1 | |
| BC034758.1 | 2 | 111 | AAH34758.1 | |
| BT019334.1 | AAV38141.1 | |||
| CR541940.1 | CAG46738.1 | |||
| CR541965.1 | CAG46763.1 | |||
| DA403369.1 | 2 | 354 | ||
| U48263.1 | 2 | 269 | AAC50651.1 | |
| X97370.1 | 1 | 81 | CAA66040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6134 | Chr.8:28294984 - 28330125 on Build GRCh38 | Insertion |
|
| dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map