Assay Details
Target Gene Details
Entrez Gene ID: | 54904 |
Gene Name: | Wolf-Hirschhorn syndrome candidate 1-like 1 |
Gene Aliases: |
NSD3, WHISTLE, pp14328 |
Location: |
Chr.8:38275042-38382272 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WHSC1L1 | NM_023034.1 | 23 | 4516 | NP_075447.1 |
| AF332469.1 | 23 | 4516 | AAK00355.1 | |
| AJ295990.1 | 23 | 4311 | CAC28350.1 | |
| AJ295991.1 | 22 | 4164 | CAC28351.1 | |
| AL832895.1 | 3 | 909 | ||
| BC101717.1 | 23 | 4090 | AAI01718.1 | |
| BC113469.1 | 23 | 4090 | ||
| BC115006.1 | 23 | 4111 | AAI15007.1 | |
| BC143510.1 | 23 | 4057 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7164n100 | Chr.8:38194164 - 38310837 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map