Assay Details
Target Gene Details
Entrez Gene ID: | 4824 |
Gene Name: | NK3 homeobox 1 |
Gene Aliases: |
BAPX2, NKX3, NKX3.1, NKX3A |
Location: |
Chr.8:23678693-23682937 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX3-1 | NM_001256339.1 | 3 | 341 | NP_001243268.1 |
| NM_006167.3 | 2 | 614 | NP_006158.2 | |
| AF247704.1 | 2 | 614 | AAG09781.1 | |
| AF249669.1 | 3 | 444 | AAG39735.1 | |
| AF249670.1 | 2 | 429 | AAG39736.1 | |
| AF249671.1 | 3 | 369 | AAG39737.1 | |
| AF249672.1 | 3 | 351 | AAG39738.1 | |
| BC074863.2 | 2 | 598 | AAH74863.1 | |
| BC074864.2 | 2 | 598 | AAH74864.1 | |
| U80669.1 | 2 | 595 | AAB38747.1 | |
| U91540.1 | 2 | 566 | AAB68662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831269 | Chr.8:23661667 - 23849831 on Build GRCh38 | Gain |
|
| nsv1028341 | Chr.8:23550380 - 24287905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map