Assay Details
Target Gene Details
Entrez Gene ID: | 84301 |
Gene Name: | DNA damage inducible 1 homolog 2 |
Gene Aliases: |
- |
Location: |
Chr.1:15617458-15661057 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDI2 | NM_032341.4 | 3 | 610 | NP_115717.3 |
| AK057374.1 | ||||
| AK293006.1 | 3 | 610 | ||
| BC006011.1 | 3 | 525 | AAH06011.1 |
More Information
Set Membership: |
|

Genomic Map