Assay Details
Target Gene Details
Entrez Gene ID: | 22985 |
Gene Name: | apoptotic chromatin condensation inducer 1 |
Gene Aliases: |
ACINUS, ACN, fSAP152 |
Location: |
Chr.14:23058564-23095614 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACIN1 | NM_001164814.1 | 19 | 4636 | NP_001158286.1 |
| NM_001164815.1 | 18 | 4555 | NP_001158287.1 | |
| NM_001164816.1 | 12 | 2194 | NP_001158288.1 | |
| NM_001164817.1 | 13 | 2256 | NP_001158289.1 | |
| NM_014977.3 | 19 | 4675 | NP_055792.1 | |
| XM_005267415.3 | 19 | 4476 | XP_005267472.1 | |
| XM_005267416.3 | 19 | 4443 | XP_005267473.1 | |
| XM_005267418.1 | 12 | 2491 | XP_005267475.1 | |
| AB014570.2 | 19 | 4184 | BAA31645.2 | |
| AF124726.1 | 19 | 4674 | AAD56724.1 | |
| AF124727.1 | 13 | 2210 | AAD56725.1 | |
| AF124728.1 | 12 | 2185 | AAD56726.1 | |
| AL050382.1 | 12 | 3280 | CAB43681.2 | |
| AL833228.1 | 14 | 2525 | CAH10608.1 | |
| BX247975.1 | 12 | 2464 | CAD62309.1 |
More Information
Set Membership: |
|

Genomic Map