Assay Details
Target Gene Details
| Entrez Gene ID: | 51338 | 
| Gene Name: | membrane spanning 4-domains A4A | 
| Gene Aliases: | 4SPAN1, CD20-L1, CD20L1, HDCME31P, MS4A4, MS4A7 | 
| Location: | Chr.11:60280541-60308972 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 8 - Exon 9 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MS4A4A | NM_001243266.1 | NP_001230195.1 | ||
| NM_024021.3 | NP_076926.2 | |||
| NM_148975.2 | NP_683876.1 | |||
| XM_017017909.1 | XP_016873398.1 | |||
| AB013102.1 | BAB18738.1 | |||
| AB022821.1 | BAB61018.1 | |||
| AF068288.1 | AAF65507.1 | |||
| AF237912.1 | AAK37594.1 | |||
| AF350500.1 | AAL56220.1 | |||
| AF354928.1 | AAL08486.1 | |||
| AK075081.1 | BAC11389.1 | |||
| AK292089.1 | ||||
| AK314033.1 | ||||
| BC020648.1 | AAH20648.1 | |||
| CB046393.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1145537 | Chr.11:60299618 - 60330486 on Build GRCh38 | Deletion |  MS4A4A | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top