Assay Details
Target Gene Details
Entrez Gene ID: | 63826 |
Gene Name: | serine racemase |
Gene Aliases: |
ILV1, ISO1 |
Location: |
Chr.17:2303371-2325267 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRR | NM_001304803.1 | 6 | 624 | NP_001291732.1 |
| NM_021947.2 | 7 | 796 | NP_068766.1 | |
| XM_006721565.3 | 7 | 828 | XP_006721628.1 | |
| XM_006721566.3 | 7 | 2006 | XP_006721629.1 | |
| XM_011523974.2 | 7 | 935 | XP_011522276.1 | |
| AF169974.1 | 6 | 733 | AAG27081.1 | |
| AK023169.1 | 7 | 785 | BAB14442.1 | |
| AK223120.1 | 7 | 789 | BAD96840.1 | |
| AL834378.1 | 5 | 561 | CAD39041.1 | |
| AY034081.1 | 6 | 717 | AAK58495.1 | |
| BC074728.2 | 6 | 735 | AAH74728.1 | |
| CR457300.1 | 6 | 717 | CAG33581.1 | |
| DA297969.1 |
Target Gene Details
Entrez Gene ID: | 55720 |
Gene Name: | TSR1, ribosome maturation factor |
Gene Aliases: |
- |
Location: |
Chr.17:2322678-2337384 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSR1 | NM_018128.4 | 15 | 4310 | NP_060598.3 |
| AK026565.1 | 15 | 3383 | ||
| BC019090.2 | 14 | 3218 | AAH19090.2 | |
| BC110851.1 | 15 | 3888 | AAI10852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain |
|
| nsv1067416 | Chr.17:2319186 - 2343258 on Build GRCh38 | Loss |
|
| nsv469765 | Chr.17:2213155 - 2400446 on Build GRCh38 | Loss |
|
| nsv833336 | Chr.17:2176922 - 2400456 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map