Assay Details
Target Gene Details
Entrez Gene ID: | 58985 |
Gene Name: | interleukin 22 receptor subunit alpha 1 |
Gene Aliases: |
CRF2-9, IL22R, IL22R1 |
Location: |
Chr.1:24119771-24143285 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL22RA1 | NM_021258.3 | 2 | 300 | NP_067081.2 |
| XM_011541882.1 | 2 | 194 | XP_011540184.1 | |
| AF286095.1 | 2 | 120 | AAG22073.1 | |
| AK226001.1 | 2 | 134 | ||
| AK292204.1 | 2 | 195 | ||
| AK308522.1 | 2 | 143 | ||
| AK313971.1 | 2 | 300 | ||
| BC029273.1 | 2 | 136 | AAH29273.1 |
More Information
Set Membership: |
|

Genomic Map