Assay Details
Target Gene Details
Entrez Gene ID: | 84892 |
Gene Name: | protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-) |
Gene Aliases: |
AGO61, C3orf39, GTDC2, MDDGA8 |
Location: |
Chr.3:43079229-43106092 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMGNT2 | NM_032806.5 | 2 | 1241 | NP_116195.2 |
| XM_005265515.2 | 3 | 1372 | XP_005265572.1 | |
| XM_011534163.2 | 3 | 1360 | XP_011532465.1 | |
| XM_011534164.1 | 2 | 1942 | XP_011532466.1 | |
| XM_017007353.1 | 4 | 1451 | XP_016862842.1 | |
| AK027472.1 | 1 | 501 | BAB55137.1 | |
| AK092147.1 | 2 | 1029 | BAC03816.1 | |
| AK124737.1 | 1 | 2209 | ||
| BC060861.1 | 2 | 1231 | AAH60861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954473 | Chr.3:43076109 - 43082208 on Build GRCh38 | Deletion |
|
| nsv590149 | Chr.3:42926105 - 43133715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map