Assay Details
Target Gene Details
Entrez Gene ID: | 6470 |
Gene Name: | serine hydroxymethyltransferase 1 |
Gene Aliases: |
CSHMT, SHMT |
Location: |
Chr.17:18327860-18363563 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHMT1 | NM_001281786.1 | NP_001268715.1 | ||
| NM_004169.4 | NP_004160.3 | |||
| NM_148918.2 | NP_683718.1 | |||
| XM_005256767.3 | XP_005256824.1 | |||
| XM_011523992.2 | XP_011522294.1 | |||
| XM_017024957.1 | XP_016880446.1 | |||
| XM_017024958.1 | XP_016880447.1 | |||
| XM_017024959.1 | XP_016880448.1 | |||
| XM_017024960.1 | XP_016880449.1 | |||
| AK223552.1 | BAD97272.1 | |||
| AK298415.1 | ||||
| BC007979.2 | AAH07979.1 | |||
| BC022874.1 | AAH22874.1 | |||
| BC038598.1 | AAH38598.1 | |||
| L11931.1 | AAA63257.1 | |||
| L23928.1 | AAA36018.1 | |||
| Y14485.1 | CAB54838.1 | |||
| Y14486.1 | CAB54839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509653 | Chr.17:18328532 - 18406188 on Build GRCh38 | Insertion |
|
| nsv833386 | Chr.17:18266947 - 18429412 on Build GRCh38 | Gain |
|
| esv2758677 | Chr.17:18331211 - 18543070 on Build GRCh38 | Gain+Loss |
|
| nsv428333 | Chr.17:18296767 - 18543070 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map