Assay Details
Target Gene Details
Entrez Gene ID: | 220296 |
Gene Name: | hepatic and glial cell adhesion molecule |
Gene Aliases: |
GlialCAM, MLC2A, MLC2B |
Location: |
Chr.11:124919244-124936412 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEPACAM | NM_152722.4 | 7 | 1617 | NP_689935.2 |
| XM_005271449.2 | 7 | 1614 | XP_005271506.1 | |
| XM_017017361.1 | 7 | 1773 | XP_016872850.1 | |
| AK098396.1 | 7 | 1255 | BAC05297.1 | |
| AK122595.1 | BAC85486.1 | |||
| AL834419.1 | 4 | 459 | CAD39081.1 | |
| AY047587.1 | 7 | 1255 | AAQ93018.1 | |
| BC104831.1 | 7 | 1239 | AAI04832.1 | |
| BC113562.1 | 7 | 1239 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745189 | Chr.11:124911992 - 125036537 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map