Assay Details
Target Gene Details
Entrez Gene ID: | 5554 |
Gene Name: | proline rich protein HaeIII subfamily 1 |
Gene Aliases: |
Db-s, PA, PIF-S, PRH2, Pr1/Pr2 |
Location: |
Chr.12:10880961-11171623 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1 | NM_001291314.1 | NP_001278243.1 | ||
| NM_001291315.1 | NP_001278244.1 | |||
| AB593029.1 | ||||
| BI772091.1 | ||||
| BM476484.1 | ||||
| BU543118.1 |
Target Gene Details
Entrez Gene ID: | 100533464 |
Gene Name: | PRH1-PRR4 readthrough |
Gene Aliases: |
PRH1 |
Location: |
Chr.12:10845849-11171625 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1-PRR4 | NR_037918.2 | |||
| AB593030.1 |
Target Gene Details
Entrez Gene ID: | 50838 |
Gene Name: | taste 2 receptor member 13 |
Gene Aliases: |
T2R13, TRB3 |
Location: |
Chr.12:10907926-10909562 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS2R13 | NM_023920.2 | NP_076409.1 | ||
| BC095518.1 | AAH95518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map