Assay Details
Target Gene Details
Entrez Gene ID: | 387129 |
Gene Name: | neuropeptide S receptor 1 |
Gene Aliases: |
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1 |
Location: |
Chr.7:34658239-34878332 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPSR1 | NM_001300933.1 | NP_001287862.1 | ||
| NM_001300934.1 | NP_001287863.1 | |||
| NM_001300935.1 | NP_001287864.1 | |||
| NM_207172.1 | NP_997055.1 | |||
| NM_207173.1 | NP_997056.1 | |||
| AK172847.1 | BAD18811.1 | |||
| AY310326.1 | AAQ76966.1 | |||
| AY310327.1 | AAQ76967.1 | |||
| AY310328.1 | AAQ76968.1 | |||
| AY310330.1 | AAQ76970.1 | |||
| AY310331.1 | AAQ76971.1 | |||
| AY310332.1 | AAQ76972.1 | |||
| AY956446.1 | AAX63744.1 | |||
| AY956447.1 | AAX63745.1 | |||
| AY956448.1 | AAX63746.1 | |||
| BC132693.1 | ||||
| BC132695.1 | ||||
| BC144380.1 |
Target Gene Details
Entrez Gene ID: | 404744 |
Gene Name: | NPSR1 antisense RNA 1 |
Gene Aliases: |
AAA1 |
Location: |
Chr.7:34346512-34834331 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPSR1-AS1 | NR_015356.2 | |||
| NR_033664.1 | ||||
| NR_033665.1 | ||||
| AY312364.1 | AAQ85133.1 | |||
| AY312365.1 | ||||
| AY312366.1 | AAQ85134.1 | |||
| AY312367.1 | AAQ85135.1 | |||
| AY312368.1 | AAQ85136.1 | |||
| AY312369.1 | AAQ85137.1 | |||
| AY312370.1 | AAQ85138.1 | |||
| AY312371.1 | AAQ85139.1 | |||
| AY312372.1 | AAQ85140.1 | |||
| AY312373.1 | AAQ85141.1 | |||
| AY321515.1 | AAQ87609.1 | |||
| AY321516.1 | AAQ87610.1 | |||
| BX649082.1 | ||||
| U85653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map