Assay Details
Target Gene Details
Entrez Gene ID: | 2191 |
Gene Name: | fibroblast activation protein alpha |
Gene Aliases: |
DPPIV, FAPA, SIMP |
Location: |
Chr.2:162170684-162243535 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAP | NM_001291807.1 | NP_001278736.1 | ||
| NM_004460.3 | NP_004451.2 | |||
| XM_011510796.2 | XP_011509098.1 | |||
| AK055327.1 | ||||
| AK297118.1 | ||||
| AK310293.1 | ||||
| AL832166.1 | ||||
| BC026250.1 | AAH26250.1 | |||
| U09278.1 | AAB49652.1 |
Target Gene Details
Entrez Gene ID: | 101929532 |
Gene Name: | uncharacterized LOC101929532 |
Gene Aliases: |
- |
Location: |
Chr.2:162114441-162172739 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929532 | NR_110255.1 | 4 | 1619 | |
| BX648358.1 | 4 | 1619 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834440 | Chr.2:162145758 - 162318828 on Build GRCh38 | Gain |
|
| nsv834439 | Chr.2:162057118 - 162223456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map