Assay Details
Target Gene Details
Entrez Gene ID: | 51460 |
Gene Name: | Scm-like with four mbt domains 1 |
Gene Aliases: |
RU1, SFMBT, hSFMBT |
Location: |
Chr.3:52899209-53046655 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFMBT1 | NM_016329.3 | 20 | 2796 | NP_057413.2 |
| XM_005265221.3 | 20 | 2912 | XP_005265278.1 | |
| XM_006713203.3 | 21 | 3034 | XP_006713266.1 | |
| XM_006713204.3 | 20 | 2734 | XP_006713267.1 | |
| XM_011533824.2 | 21 | 2910 | XP_011532126.1 | |
| XM_011533825.2 | 21 | 2757 | XP_011532127.1 | |
| AB189472.1 | 19 | 2420 | BAE43835.1 | |
| AF168132.1 | 20 | 2733 | AAF19794.1 | |
| AK313965.1 | 20 | 2791 | ||
| AL080140.1 | CAB45734.1 | |||
| BC014614.2 | 20 | 2680 | AAH14614.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map