Assay Details
Target Gene Details
Entrez Gene ID: | 2913 |
Gene Name: | glutamate metabotropic receptor 3 |
Gene Aliases: |
GLUR3, GPRC1C, MGLUR3, mGlu3 |
Location: |
Chr.7:86643914-86864879 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM3 | NM_000840.2 | NP_000831.2 | ||
| BC041407.1 | AAH41407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831053 | Chr.7:86638855 - 86811683 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map