Assay Details
Target Gene Details
Entrez Gene ID: | 81887 |
Gene Name: | LAS1 like, ribosome biogenesis factor |
Gene Aliases: |
Las1-like, dJ475B7.2 |
Location: |
Chr.X:65512582-65534806 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAS1L | NM_001170649.1 | 12 | 2068 | NP_001164120.1 |
| NM_001170650.1 | 11 | 1942 | NP_001164121.1 | |
| NM_031206.4 | 13 | 2119 | NP_112483.1 | |
| XM_005262301.1 | 13 | 2101 | XP_005262358.1 | |
| XM_011531045.1 | 12 | 1968 | XP_011529347.1 | |
| XM_017029877.1 | 12 | 2050 | XP_016885366.1 | |
| XM_017029879.1 | 12 | 1994 | XP_016885368.1 | |
| XM_017029880.1 | 11 | 1943 | XP_016885369.1 | |
| AF116730.1 | 13 | 2057 | AAO15306.1 | |
| AK022587.1 | 13 | 2069 | BAB14114.1 | |
| AK054817.1 | 3 | 1735 | ||
| AK074087.1 | 12 | 4836 | BAB84913.1 | |
| AK097383.1 | 11 | 1892 | ||
| AK125760.1 | 2 | 1596 | ||
| AK127207.1 | 2 | 1124 | ||
| AK225059.1 | 12 | 2001 | ||
| BC014545.1 | 13 | 2103 | AAH14545.1 | |
| BC018610.1 | 12 | 2016 | AAH18610.1 | |
| BC019302.1 | 12 | 2013 | AAH19302.1 |
More Information
Set Membership: |
|

Genomic Map