Assay Details
Target Gene Details
Entrez Gene ID: | 9002 |
Gene Name: | F2R like thrombin/trypsin receptor 3 |
Gene Aliases: |
PAR4 |
Location: |
Chr.19:16889015-16892019 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2RL3 | NM_003950.2 | 2 | 1226 | NP_003941.2 |
| XM_005260139.2 | 2 | 1047 | XP_005260196.1 | |
| AF055917.1 | 2 | 1226 | AAC25699.1 | |
| AF080214.1 | 2 | 1053 | AAC28860.1 | |
| AY431102.1 | 2 | 1051 | AAR08487.1 | |
| BC074782.2 | 2 | 1070 | AAH74782.2 |
More Information
Set Membership: |
|

Genomic Map