Assay Details
Target Gene Details
Entrez Gene ID: | 9575 |
Gene Name: | clock circadian regulator |
Gene Aliases: |
KAT13D, bHLHe8 |
Location: |
Chr.4:55427901-55547138 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLOCK | NM_001267843.1 | 24 | 3340 | NP_001254772.1 |
| NM_004898.3 | 23 | 2831 | NP_004889.1 | |
| XM_005265787.2 | 23 | 2877 | XP_005265844.1 | |
| XM_011534409.2 | 23 | 3286 | XP_011532711.1 | |
| XM_011534410.2 | 22 | 2668 | XP_011532712.1 | |
| XM_011534411.2 | 22 | 2627 | XP_011532713.1 | |
| XM_017008854.1 | 24 | 3217 | XP_016864343.1 | |
| XM_017008855.1 | 20 | 2426 | XP_016864344.1 | |
| AB002332.1 | 22 | 2665 | ||
| AF011568.1 | 23 | 2771 | AAB83969.1 | |
| AK223549.1 | 23 | 2771 | BAD97269.1 | |
| AK291708.1 | 23 | 2770 | ||
| BC041878.1 | 24 | 3340 | AAH41878.1 | |
| BC126157.1 | 23 | 2747 | ||
| BC126159.1 | 23 | 2747 | ||
| BX537616.1 | 7 | 866 |
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM165 | XM_011534394.2 | XP_011532696.1 |
More Information
Set Membership: |
|

Genomic Map