Assay Details
Target Gene Details
Entrez Gene ID: | 171568 |
Gene Name: | RNA polymerase III subunit H |
Gene Aliases: |
RPC22.9, RPC8 |
Location: |
Chr.22:41525799-41544606 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3H | NM_001018050.3 | NP_001018060.1 | ||
| NM_001018052.3 | NP_001018062.1 | |||
| NM_001282884.1 | NP_001269813.1 | |||
| NM_001282885.1 | NP_001269814.1 | |||
| NM_138338.4 | NP_612211.1 | |||
| XM_011529993.1 | XP_011528295.1 | |||
| XM_017028641.1 | XP_016884130.1 | |||
| AB051452.1 | ||||
| AK026524.1 | ||||
| AY092087.1 | AAM18217.1 | |||
| BC017248.2 | AAH17248.1 | |||
| BC088367.1 | AAH88367.1 | |||
| BI754673.1 | ||||
| BU173504.1 | ||||
| BX331353.2 | ||||
| BX367405.2 | ||||
| CR456459.1 | CAG30345.1 | |||
| DA449644.1 | ||||
| DA504810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
|
| nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| esv3647799 | Chr.22:41511654 - 41543254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map