Assay Details
Target Gene Details
Entrez Gene ID: | 468 |
Gene Name: | activating transcription factor 4 |
Gene Aliases: |
CREB-2, CREB2, TAXREB67, TXREB |
Location: |
Chr.22:39519709-39522686 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF4 | NM_001675.4 | NP_001666.2 | ||
| NM_182810.2 | NP_877962.1 | |||
| XM_017028807.1 | 2 | 495 | XP_016884296.1 | |
| AK057751.1 | ||||
| AK296028.1 | ||||
| BC008090.1 | AAH08090.1 | |||
| BC011994.1 | AAH11994.1 | |||
| BC016855.2 | AAH16855.1 | |||
| BC022088.2 | AAH22088.1 | |||
| BC024775.1 | AAH24775.1 | |||
| BC044895.1 | AAH44895.1 | |||
| BC073754.1 | AAH73754.1 | |||
| D90209.1 | BAA14234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834199 | Chr.22:39324389 - 39531343 on Build GRCh38 | Loss |
|
| nsv3629 | Chr.22:39504067 - 39534583 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map