Assay Details
Target Gene Details
| Entrez Gene ID: | 2475 | 
| Gene Name: | mechanistic target of rapamycin | 
| Gene Aliases: | FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS | 
| Location: | Chr.1:11106531-11262551 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 59 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MTOR | NM_004958.3 | 58 | 8154 | NP_004949.1 | 
| XM_005263438.2 | 58 | 8161 | XP_005263495.1 | |
| XM_017000900.1 | 55 | 7617 | XP_016856389.1 | |
| XM_017000901.1 | 56 | 7905 | XP_016856390.1 | |
| AB209995.1 | 58 | 8135 | ||
| AK126762.1 | 20 | 3450 | ||
| AK302863.1 | ||||
| AK304273.1 | ||||
| BC117166.1 | 57 | 8051 | ||
| U88966.1 | 58 | 8109 | AAC39933.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |  MASP2  MTOR-AS1  UBIAD1  TARDBP  ANGPTL7  EXOSC10  SRM  LOC105376736  MTOR | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top