Assay Details
Target Gene Details
Entrez Gene ID: | 54567 |
Gene Name: | delta like canonical Notch ligand 4 |
Gene Aliases: |
AOS6, hdelta2 |
Location: |
Chr.15:40929333-40939060 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLL4 | NM_019074.3 | 2 | 599 | NP_061947.1 |
| AB036931.1 | 2 | 539 | BAB16085.1 | |
| AB043894.1 | 2 | 263 | BAB18581.1 | |
| AF253468.1 | 2 | 263 | AAF76427.1 | |
| AF279305.1 | 1 | 169 | AAF81912.1 | |
| AK308755.1 | 2 | 592 | ||
| AK313831.1 | 2 | 594 | ||
| AY358894.1 | 2 | 298 | AAQ89253.1 | |
| BC106950.2 | 2 | 399 | AAI06951.1 |
More Information
Set Membership: |
|

Genomic Map