Assay Details
Target Gene Details
Entrez Gene ID: | 57223 |
Gene Name: | protein phosphatase 4 regulatory subunit 3B |
Gene Aliases: |
FLFL2, PP4R3B, PSY2, SMEK2, smk1 |
Location: |
Chr.2:55547292-55617757 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R3B | NM_001122964.2 | 17 | 3159 | NP_001116436.2 |
| NM_001282850.1 | 16 | 3063 | NP_001269779.1 | |
| NM_020463.3 | 15 | 2904 | NP_065196.1 | |
| XM_005264442.2 | 17 | 3138 | XP_005264499.1 | |
| XM_005264444.2 | 16 | 3000 | XP_005264501.1 | |
| XM_005264445.2 | 16 | 2979 | XP_005264502.1 | |
| XM_005264446.2 | 15 | 2883 | XP_005264503.1 | |
| XM_005264447.2 | 16 | 2535 | XP_005264504.1 | |
| XM_017004531.1 | 16 | 3042 | XP_016860020.1 | |
| XM_017004532.1 | 16 | 2514 | XP_016860021.1 | |
| XM_017004533.1 | 15 | 2439 | XP_016860022.1 | |
| XM_017004534.1 | 15 | 2418 | XP_016860023.1 | |
| XM_017004535.1 | 14 | 2280 | XP_016860024.1 | |
| XM_017004536.1 | 14 | 2259 | XP_016860025.1 | |
| XM_017004537.1 | 18 | 3350 | XP_016860026.1 | |
| XM_017004538.1 | 18 | 3262 | XP_016860027.1 | |
| XM_017004539.1 | 17 | 3146 | XP_016860028.1 | |
| XM_017004540.1 | 17 | 3145 | XP_016860029.1 | |
| AB037808.1 | 17 | 3022 | ||
| AK023148.1 | 6 | 1052 | BAB14430.1 | |
| AK026437.1 | 12 | 2161 | ||
| AK098321.1 | 1 | 978 | ||
| AL122106.1 | 2 | 357 | ||
| AL136556.1 | 7 | 1152 | CAB66491.2 | |
| BC006215.2 | 16 | 2934 | AAH06215.1 | |
| BC060855.1 | 15 | 2793 | AAH60855.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575244 | Chr.2:55522427 - 56940881 on Build GRCh38 | Gain |
|
| esv2759050 | Chr.2:55520103 - 55753939 on Build GRCh38 | Gain |
|
| nsv1160801 | Chr.2:55524659 - 55588556 on Build GRCh38 | Duplication |
|
| nsv520581 | Chr.2:55395689 - 55675881 on Build GRCh38 | Loss |
|
| nsv525434 | Chr.2:55489514 - 55614504 on Build GRCh38 | Loss |
|
| nsv834080 | Chr.2:55513900 - 55700793 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map