Assay Details
Target Gene Details
Entrez Gene ID: | 8013 |
Gene Name: | nuclear receptor subfamily 4 group A member 3 |
Gene Aliases: |
CHN, CSMF, MINOR, NOR1, TEC |
Location: |
Chr.9:99821855-99866893 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR4A3 | NM_006981.3 | 5 | 1899 | NP_008912.2 |
| NM_173199.2 | 5 | 1899 | NP_775291.1 | |
| NM_173200.2 | 6 | 1971 | NP_775292.1 | |
| XM_017015162.1 | 4 | 1831 | XP_016870651.1 | |
| AK226108.1 | 5 | 1881 | ||
| BX649089.1 | 2 | 267 | CAH10551.1 | |
| D78579.1 | 5 | 1901 | BAA11419.1 | |
| D85242.1 | 3 | 270 | BAA31221.1 | |
| S81243.1 | 5 | 1786 | AAB36006.1 | |
| U12767.1 | 4 | 1262 | AAB02581.1 | |
| X89894.1 | 5 | 1857 | CAA61984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528025 | Chr.9:99778104 - 99851390 on Build GRCh38 | Gain |
|
| nsv831668 | Chr.9:99805887 - 99985011 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map