Assay Details
Target Gene Details
Entrez Gene ID: | 3182 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A/B |
Gene Aliases: |
ABBP1, HNRPAB |
Location: |
Chr.5:178204507-178211183 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPAB | NM_004499.3 | NP_004490.2 | ||
| NM_031266.2 | NP_112556.2 | |||
| AK054600.1 | ||||
| AK097657.1 | BAC05134.1 | |||
| AK223425.1 | BAD97145.1 | |||
| BC001616.1 | AAH01616.1 | |||
| BC002625.2 | AAH02625.1 | |||
| BC004561.2 | AAH04561.1 | |||
| BC009359.2 | AAH09359.1 | |||
| BC036708.2 | AAH36708.1 | |||
| M65028.1 | AAA36575.1 | |||
| U76713.1 | AAC50956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019404 | Chr.5:178080079 - 178432088 on Build GRCh38 | Gain |
|
| nsv820182 | Chr.5:178203925 - 178207190 on Build GRCh38 | Gain |
|
| nsv1017664 | Chr.5:178151193 - 178206115 on Build GRCh38 | Gain |
|
| dgv1679e212 | Chr.5:178197037 - 178208373 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map