Assay Details
Target Gene Details
Entrez Gene ID: | 10574 |
Gene Name: | chaperonin containing TCP1 subunit 7 |
Gene Aliases: |
CCTETA, CCTH, NIP7-1, TCP1ETA |
Location: |
Chr.2:73233431-73253022 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT7 | NM_001009570.2 | NP_001009570.1 | ||
| NM_001166284.1 | NP_001159756.1 | |||
| NM_001166285.1 | NP_001159757.1 | |||
| NM_006429.3 | NP_006420.1 | |||
| NR_029402.1 | ||||
| NR_029403.1 | ||||
| XM_011532478.2 | XP_011530780.1 | |||
| XM_011532479.1 | XP_011530781.1 | |||
| AF026292.1 | AAC96011.1 | |||
| AK098549.1 | ||||
| AK098825.1 | ||||
| AK222478.1 | BAD96198.1 | |||
| AK291961.1 | ||||
| AK293175.1 | ||||
| AK293597.1 | ||||
| AK297846.1 | ||||
| AK298153.1 | ||||
| AK302078.1 | ||||
| BC019296.1 | AAH19296.1 | |||
| BC088351.1 | AAH88351.1 | |||
| CR456719.1 | CAG33000.1 | |||
| CR536511.1 | CAG38749.1 | |||
| U83843.1 | AAB41437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2776 | Chr.2:73204726 - 73255176 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map