Assay Details
Target Gene Details
Entrez Gene ID: | 10210 |
Gene Name: | TOP1 binding arginine/serine rich protein |
Gene Aliases: |
LUN, P53BP3, RP31, TP53BPL |
Location: |
Chr.9:32540544-32552628 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPORS | NM_001195622.1 | 1 | 70 | NP_001182551.1 |
| NM_005802.4 | 1 | 70 | NP_005793.2 | |
| AB045732.1 | BAB03714.1 | |||
| AB045733.1 | BAB03715.1 | |||
| AF098300.1 | AAD23379.1 | |||
| AU280552.1 | ||||
| BC060884.1 | AAH60884.1 | |||
| DA238762.1 | 1 | 70 |
Target Gene Details
Entrez Gene ID: | 100129250 |
Gene Name: | TOPORS antisense RNA 1 |
Gene Aliases: |
C9orf133 |
Location: |
Chr.9:32551144-32566860 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPORS-AS1 | NR_033991.1 | |||
| NR_033992.2 | ||||
| NR_102376.1 | ||||
| NR_102377.1 | ||||
| BC013655.1 | 1 | 214 | AAH13655.1 | |
| BI916035.1 | ||||
| CB048380.1 | ||||
| DA702677.1 | ||||
| DA753662.1 | ||||
| HY032383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map