Assay Details
Target Gene Details
Entrez Gene ID: | 84296 |
Gene Name: | GINS complex subunit 4 |
Gene Aliases: |
SLD5 |
Location: |
Chr.8:41529206-41545046 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS4 | NM_032336.2 | 1 | 74 | NP_115712.1 |
| XM_005273659.3 | XP_005273716.1 | |||
| AK095334.1 | ||||
| AK299207.1 | 1 | 62 | ||
| AK313373.1 | ||||
| BC005995.1 | 1 | 53 | AAH05995.1 | |
| BC027454.1 | 1 | 59 | AAH27454.1 | |
| DC376966.1 | 1 | 74 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss |
|
| esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map