Assay Details
Target Gene Details
Entrez Gene ID: | 4889 |
Gene Name: | neuropeptide Y receptor Y5 |
Gene Aliases: |
NPY5-R, NPYR5, NPYY5-R |
Location: |
Chr.4:163336968-163352277 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY5R | NM_001317091.1 | 4 | 2628 | NP_001304020.1 |
| NM_001317092.1 | 5 | 1508 | NP_001304021.1 | |
| NM_006174.3 | 4 | 1380 | NP_006165.1 | |
| XM_005263038.3 | 4 | 8262 | XP_005263095.1 | |
| XM_011532015.2 | 5 | 8299 | XP_011530317.1 | |
| XM_011532017.2 | 5 | 8390 | XP_011530319.1 | |
| XM_017008255.1 | 5 | 1340 | XP_016863744.1 | |
| XM_017008256.1 | 6 | 8427 | XP_016863745.1 | |
| BC042416.1 | 4 | 1288 | AAH42416.1 | |
| U56079.1 | 1 | 1161 | AAC50623.1 | |
| U66275.1 | 1 | 1123 | AAC50741.1 | |
| U94320.1 | 2 | 1176 | AAC51295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv596023 | Chr.4:163349101 - 163379624 on Build GRCh38 | Loss |
|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map