Assay Details
Target Gene Details
Entrez Gene ID: | 6309 |
Gene Name: | sterol-C5-desaturase |
Gene Aliases: |
ERG3, S5DES, SC5DL |
Location: |
Chr.11:121292679-121313410 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SC5D | NM_001024956.2 | 1 | 90 | NP_001020127.1 |
| NM_006918.4 | NP_008849.2 | |||
| AF069469.1 | ||||
| AK096847.1 | ||||
| AK222686.1 | BAD96406.1 | |||
| AK223141.1 | BAD96861.1 | |||
| AK312634.1 | ||||
| BC012333.1 | 1 | 58 | AAH12333.1 | |
| BC050427.1 | AAH50427.1 | |||
| BI559520.1 |
More Information
Set Membership: |
|

Genomic Map