Assay Details
Target Gene Details
Entrez Gene ID: | 5020 |
Gene Name: | oxytocin/neurophysin I prepropeptide |
Gene Aliases: |
OT, OT-NPI, OXT-NPI |
Location: |
Chr.20:3068871-3072517 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXT | NM_000915.3 | 1 | 85 | NP_000906.1 |
| XM_011529238.1 | 2 | 222 | XP_011527540.1 | |
| BC069144.1 | 1 | 85 | AAH69144.1 | |
| BC101841.1 | 1 | 83 | AAI01842.1 | |
| BC101843.1 | 1 | 83 | AAI01844.1 | |
| KJ534910.1 | 1 | 49 | ||
| M25650.1 | 1 | 66 | AAA59977.1 | |
| M62611.1 | AAA59979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
| nsv523746 | Chr.20:3069244 - 3072609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map